Assay Details
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172493.2 | NP_766081.1 | 23 - 24 | 3081 | 84 | |
GenBank mRNA | AB713187.1 | - | 24 - 25 | 2876 | 84 | |
AK083254.1 | - | 16 - 17 | 2013 | 84 | ||
AK088648.1 | - | 23 - 24 | 3015 | 84 | ||
AY312280.1 | - | 23 - 24 | 3018 | 84 | ||
Y15910.1 | - | 4 - 5 | 588 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
