Assay Details
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172493.2 | NP_766081.1 | 6 - 7 | 907 | 75 | |
GenBank mRNA | AB713187.1 | - | 7 - 8 | 702 | 75 | |
AK088648.1 | - | 6 - 7 | 841 | 75 | ||
AK153986.1 | - | 6 - 7 | 895 | 75 | ||
AY312280.1 | - | 6 - 7 | 844 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
