| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00012328.mm.1 |
NM_001013387.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012328.mm.1 |
NM_001013387.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012328.mm.1 |
NM_001111076.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012328.mm.1 |
NM_001111076.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012328.mm.1 |
TC0X00001937.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012332.mm.1 |
NM_001013387.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012332.mm.1 |
NM_001013387.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012332.mm.1 |
NM_001111076.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012332.mm.1 |
NM_001111076.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012332.mm.1 |
TC0X00001937.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012319.mm.1 |
NM_001013387.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012319.mm.1 |
NM_001013387.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012319.mm.1 |
NM_001111076.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012319.mm.1 |
NM_001111076.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012319.mm.1 |
TC0X00001937.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00012330.mm.1 |
NM_001013387.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012330.mm.1 |
NM_001013387.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012330.mm.1 |
NM_001111076.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012330.mm.1 |
NM_001111076.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00012330.mm.1 |
TC0X00001937.mm.1 |
MTA TC |