Assay Details
| Gene Symbol: Zfp182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013387.2 | NP_001013405.2 | 3 - 4 | 317 | 92 | |
| NM_001111076.1 | NP_001104546.1 | 3 - 4 | 341 | 92 | ||
| GenBank mRNA | AK155039.1 | - | 3 - 4 | 289 | 92 | |
| BC063066.1 | - | 3 - 4 | 282 | 6822685 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp182
Genomic Map