Assay Details
Gene Symbol: Rp2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110966.1 | 5 - 6 | 1245 | 85 | ||
GenBank mRNA | AK044002.1 | - | 5 - 6 | 1152 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
