Assay Details
Gene Symbol: Rp2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110967.1 | 4 - 5 | 1212 | 111 | ||
GenBank mRNA | AK076907.1 | - | 4 - 5 | 1074 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
