Assay Details
| Gene Symbol: Nt5c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252374.1 | NP_001239303.1 | 2 - 3 | 102 | 116 | |
| GenBank mRNA | AK160139.1 | - | 2 - 3 | 102 | 116 | |
| BC038029.1 | - | 2 - 3 | 95 | 4018956 | 116 | |
| CB571852.1 | - | 2 - 3 | 107 | 116 | ||
| CX734167.1 | - | 7 - 8 | 116 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c3
Genomic Map