Assay Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009593.2 | NP_033723.1 | 11 - 12 | 1512 | 68 | |
| GenBank mRNA | CV558133.1 | - | 3 - 4 | 323 | 68 | |
More Information
Set Membership: |
|

Abcg1
Genomic Map