Assay Details
| Gene Symbol: Pde9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163748.1 | NP_001157220.1 | 9 - 10 | 685 | 72 | |
| NM_008804.4 | NP_032830.3 | 9 - 10 | 685 | 72 | ||
| GenBank mRNA | AF031147.1 | - | 9 - 10 | 640 | 72 | |
| AF068247.1 | - | 9 - 10 | 649 | 72 | ||
| AK030509.1 | - | 9 - 10 | 669 | 72 | ||
| AK168143.1 | - | 9 - 10 | 667 | 72 | ||
| BC025882.1 | - | 9 - 10 | 643 | 72 | ||
| BC038675.1 | - | 9 - 10 | 72 | |||
| BC046915.1 | - | 9 - 10 | 634 | 72 | ||
| BC061163.1 | - | 9 - 10 | 665 | 6533665 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde9a
Genomic Map