Assay Details
| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039365.2 | NP_001034454.1 | 1 - 2 | 61 | 64 | |
| NM_008614.2 | NP_032640.1 | 1 - 2 | 61 | 64 | ||
| GenBank mRNA | AK013799.1 | - | 1 - 2 | 62 | 64 | |
| AK019323.1 | - | 1 - 2 | 62 | 64 | ||
| AK028057.1 | - | 1 - 2 | 61 | 64 | ||
| AK083103.1 | - | 1 - 2 | 62 | 64 | ||
| AK140055.1 | - | 1 - 2 | 62 | 64 | ||
| BC048492.1 | - | 1 - 2 | 61 | 6593408 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013799.1, AK019323.1, AK028057.1, AK083103.1, AK140055.1, BC048492.1, NM_001039365.2, NM_008614.2 transcripts.
Set Membership: |
|

Mobp
Genomic Map