Assay Details
| Gene Symbol: Shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290684.1 | NP_001277613.1 | 8 - 9 | 3942 | 63 | |
| NM_001290685.1 | NP_001277614.1 | 4 - 5 | 1267 | 63 | ||
| NM_001290686.1 | NP_001277615.1 | 4 - 5 | 1074 | 63 | ||
| NM_001290687.1 | NP_001277616.1 | 4 - 5 | 1256 | 63 | ||
| NM_172441.3 | NP_766029.2 | 8 - 9 | 3960 | 63 | ||
| GenBank mRNA | AK029338.1 | - | 4 - 5 | 948 | 63 | |
| AK032256.1 | - | 4 - 5 | 949 | 63 | ||
| AK049850.1 | - | 4 - 5 | 1006 | 63 | ||
| BC092373.1 | - | 4 - 5 | 1262 | 63 | ||
| DQ358971.1 | - | 8 - 9 | 3920 | 63 | ||
| EF071946.1 | - | 8 - 9 | 3960 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shroom2
Genomic Map