Assay Details
| Gene Symbol: Shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290684.1 | NP_001277613.1 | 7 - 8 | 3780 | 63 | |
| NM_001290685.1 | NP_001277614.1 | 3 - 4 | 1105 | 63 | ||
| NM_001290686.1 | NP_001277615.1 | 3 - 4 | 912 | 63 | ||
| NM_001290687.1 | NP_001277616.1 | 3 - 4 | 1094 | 63 | ||
| NM_172441.3 | NP_766029.2 | 7 - 8 | 3798 | 63 | ||
| GenBank mRNA | AK029338.1 | - | 3 - 4 | 786 | 63 | |
| AK032256.1 | - | 3 - 4 | 787 | 63 | ||
| AK049850.1 | - | 3 - 4 | 844 | 63 | ||
| BC092373.1 | - | 3 - 4 | 1100 | 63 | ||
| EF071946.1 | - | 7 - 8 | 3798 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shroom2
Genomic Map