Assay Details
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037761.2 | NP_001032850.1 | 8 - 9 | 858 | 100 | |
GenBank mRNA | AK153367.1 | - | 8 - 9 | 635 | 100 | |
AK153394.1 | - | 8 - 9 | 635 | 100 | ||
AK159390.1 | - | 8 - 9 | 832 | 100 | ||
AK187793.1 | - | 3 - 4 | 186 | 100 | ||
AK200176.1 | - | 3 - 4 | 186 | 100 | ||
AK219201.1 | - | 3 - 4 | 186 | 100 | ||
U10406.1 | - | 8 - 9 | 778 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
