Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400014245.mm.1 |
NM_001037761.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014245.mm.1 |
TC0400001741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014249.mm.1 |
NM_001037761.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014249.mm.1 |
NM_001271405.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014249.mm.1 |
NM_009798.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014249.mm.1 |
TC0400001741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400014248.mm.1 |
NM_001037761.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014248.mm.1 |
NM_001271405.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014248.mm.1 |
NM_009798.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400014248.mm.1 |
TC0400001741.mm.1 |
MTA TC |