Assay Details
Gene Symbol: Aldh4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175438.4 | NP_780647.3 | 4 - 5 | 539 | 65 | |
GenBank mRNA | AK044712.1 | - | 4 - 5 | 335 | 65 | |
BC024133.1 | - | 3 - 4 | 257 | 65 | ||
BC039281.1 | - | 4 - 5 | 323 | 4975491 | 65 | |
BC056226.1 | - | 4 - 5 | 423 | 5064715 | 65 | |
BC060650.1 | - | 4 - 5 | 539 | 30354228 | 65 | |
CA494657.1 | - | 4 - 5 | 576 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
