Assay Details
| Gene Symbol: Nat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168577.1 | NP_001162049.1 | 1 - 2 | 30 | 57 | |
| GenBank mRNA | AK007583.1 | - | 1 - 2 | 39 | 57 | |
| BC012972.1 | - | 1 - 2 | 48 | 3992093 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC012972.1, NM_001168577.1 transcripts.
Set Membership: |
|

Nat2
Genomic Map