Assay Details
| Gene Symbol: Lsm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015816.4 | NP_056631.2 | 3 - 4 | 368 | 77 | |
| GenBank mRNA | AJ249439.1 | - | 3 - 4 | 208 | 77 | |
| AK010910.1 | - | 3 - 4 | 204 | 77 | ||
| AK089028.1 | - | 3 - 4 | 221 | 77 | ||
| AK143470.1 | - | 3 - 4 | 192 | 77 | ||
| AK167784.1 | - | 3 - 4 | 232 | 77 | ||
| BC026747.1 | - | 3 - 4 | 351 | 4024321 | 77 | |
| CA322716.1 | - | 3 - 4 | 368 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsm4
Genomic Map