Assay Details
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133772.2 | NP_598533.2 | 7 - 8 | 628 | 95 | |
GenBank mRNA | AK004835.1 | - | 7 - 8 | 741 | 95 | |
AK154336.1 | - | 7 - 8 | 629 | 95 | ||
BC002150.1 | - | 7 - 8 | 587 | 3484896 | 95 | |
CX237617.1 | - | 6 - 7 | 410 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
