Assay Details
| Gene Symbol: Ocel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293800.1 | NP_001280729.1 | 1 - 2 | 691 | 75 | |
| GenBank mRNA | AK163762.1 | - | 692 | 75 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Recommended Assay  Mus musculus  1700  Inventoried  Probe spans exons | 
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Ocel1
Ocel1 Genomic Map
Genomic Map Back To Top
 Back To Top