Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300011275.mm.1 |
NM_145938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011275.mm.1 |
NM_145938.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011275.mm.1 |
TC1300001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011276.mm.1 |
NM_145938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011276.mm.1 |
NM_145938.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011276.mm.1 |
TC1300001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011274.mm.1 |
NM_145938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011274.mm.1 |
NM_145938.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011274.mm.1 |
TC1300001804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011277.mm.1 |
NM_145938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011277.mm.1 |
NM_145938.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011277.mm.1 |
TC1300001804.mm.1 |
MTA TC |