Assay Details
| Gene Symbol: Bicd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039179.2 | NP_001034268.1 | 2 - 3 | 582 | 57 | |
| NM_001039180.2 | NP_001034269.1 | 3 - 4 | 478 | 57 | ||
| NM_029791.4 | NP_084067.1 | 2 - 3 | 582 | 57 | ||
| GenBank mRNA | AJ250106.1 | - | 2 - 3 | 541 | 57 | |
| AK002683.1 | - | 2 - 3 | 264 | 57 | ||
| AK122348.1 | - | 2 - 3 | 555 | 57 | ||
| AK156407.1 | - | 2 - 3 | 537 | 57 | ||
| AK171300.1 | - | 3 - 4 | 436 | 57 | ||
| BC032198.1 | - | 2 - 3 | 539 | 5324113 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bicd2
Genomic Map