Assay Details
| Gene Symbol: Auh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016709.2 | NP_057918.2 | 3 - 4 | 380 | 108 | |
| GenBank mRNA | AK003219.1 | - | 3 - 4 | 359 | 108 | |
| AK019978.1 | - | 3 - 4 | 357 | 108 | ||
| AK134886.1 | - | 3 - 4 | 352 | 108 | ||
| BC026525.1 | - | 3 - 4 | 318 | 108 | ||
| BC049597.1 | - | 3 - 4 | 364 | 6593786 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Auh
Genomic Map