Assay Details
| Gene Symbol: Auh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016709.2 | NP_057918.2 | 5 - 6 | 556 | 66 | |
| GenBank mRNA | AK134886.1 | - | 5 - 6 | 528 | 66 | |
| BC026525.1 | - | 5 - 6 | 494 | 66 | ||
| BC049597.1 | - | 5 - 6 | 540 | 6593786 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Auh
Genomic Map