Assay Details
| Gene Symbol: Spock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166463.1 | NP_001159935.1 | 3 - 4 | 376 | 71 | |
| NM_001166464.1 | NP_001159936.1 | 4 - 5 | 385 | 71 | ||
| NM_001166465.1 | NP_001159937.1 | 3 - 4 | 376 | 71 | ||
| NM_001166466.1 | NP_001159938.1 | 4 - 5 | 385 | 71 | ||
| NM_009262.3 | NP_033288.2 | 4 - 5 | 385 | 71 | ||
| GenBank mRNA | AK039058.1 | - | 3 - 4 | 358 | 71 | |
| AK049019.1 | - | 3 - 4 | 344 | 71 | ||
| AK051191.1 | - | 3 - 4 | 336 | 71 | ||
| BC075625.1 | - | 4 - 5 | 328 | 6842221 | 71 | |
| BC158020.1 | - | 4 - 5 | 292 | 9007326 | 71 | |
| X92864.1 | - | 4 - 5 | 377 | 71 | ||
More Information
Set Membership: |
|

Spock1
Genomic Map