Assay Details
Gene Symbol: Spock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166463.1 | NP_001159935.1 | 4 - 5 | 488 | 71 | |
NM_001166464.1 | NP_001159936.1 | 5 - 6 | 497 | 71 | ||
NM_001166465.1 | NP_001159937.1 | 4 - 5 | 488 | 71 | ||
NM_001166466.1 | NP_001159938.1 | 5 - 6 | 497 | 71 | ||
NM_009262.3 | NP_033288.2 | 5 - 6 | 497 | 71 | ||
GenBank mRNA | AK036189.1 | - | 5 - 6 | 498 | 71 | |
AK039058.1 | - | 4 - 5 | 470 | 71 | ||
AK049019.1 | - | 4 - 5 | 456 | 71 | ||
AK051191.1 | - | 4 - 5 | 448 | 71 | ||
BB628964.1 | - | 5 - 6 | 498 | 71 | ||
BC075625.1 | - | 5 - 6 | 440 | 6842221 | 71 | |
BC158020.1 | - | 5 - 6 | 404 | 9007326 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
