Assay Details
| Gene Symbol: Pdlim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114088.2 | NP_001107560.1 | 6 - 7 | 626 | 83 | |
| GenBank mRNA | AK033091.1 | - | 618 | 83 | ||
| AK142933.1 | - | 6 - 7 | 610 | 83 | ||
| AK167476.1 | - | 6 - 7 | 618 | 83 | ||
More Information
Set Membership: |
|

Pdlim7
Genomic Map