Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800017455.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017455.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017455.mm.1 |
TC0800002313.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017458.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017458.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017458.mm.1 |
TC0800002313.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017452.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017452.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017452.mm.1 |
TC0800002313.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017457.mm.1 |
NM_025436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017457.mm.1 |
NM_025436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017457.mm.1 |
TC0800002313.mm.1 |
MTA TC |