Assay Details
Gene Symbol: Spock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252620.1 | NP_001239549.1 | 10 - 11 | 1069 | 80 | |
NM_001252621.1 | NP_001239550.1 | 9 - 10 | 1060 | 80 | ||
NM_023689.3 | NP_076178.1 | 10 - 11 | 1183 | 80 | ||
GenBank mRNA | AJ278998.1 | - | 10 - 11 | 1077 | 80 | |
AK013644.1 | - | 10 - 11 | 1112 | 80 | ||
AK049843.1 | - | 9 - 10 | 1061 | 80 | ||
AK140736.1 | - | 3 - 4 | 370 | 80 | ||
AK220280.1 | - | 10 - 11 | 1167 | 80 | ||
BC017601.1 | - | 10 - 11 | 1082 | 4502169 | 80 | |
BC053334.1 | - | 10 - 11 | 1054 | 6492113 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
