Assay Details
| Gene Symbol: Papd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169131.1 | NP_001162602.1 | 4 - 5 | 672 | 67 | |
| NM_198600.2 | NP_941002.2 | 4 - 5 | 672 | 67 | ||
| GenBank mRNA | AK152550.1 | - | 4 - 5 | 373 | 67 | |
| BC059846.1 | - | 4 - 5 | 647 | 6841188 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Papd7
Genomic Map