| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300014855.mm.1 |
NM_001169131.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014855.mm.1 |
NM_001169131.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014855.mm.1 |
NM_198600.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014855.mm.1 |
NM_198600.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014855.mm.1 |
TC1300002274.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300014854.mm.1 |
NM_001169131.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014854.mm.1 |
NM_001169131.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014854.mm.1 |
NM_198600.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014854.mm.1 |
NM_198600.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014854.mm.1 |
TC1300002274.mm.1 |
MTA TC |