Assay Details
| Gene Symbol: Papd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169131.1 | NP_001162602.1 | 9 - 10 | 1348 | 95 | |
| NM_198600.2 | NP_941002.2 | 9 - 10 | 1348 | 95 | ||
| GenBank mRNA | AK152550.1 | - | 9 - 10 | 1049 | 95 | |
| BQ180094.1 | - | 2 - 3 | 149 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papd7
Genomic Map