Assay Details
Gene Symbol: 1700001L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027035.1 | NP_081311.1 | 2 - 3 | 408 | 95 | |
GenBank mRNA | AK005609.1 | - | 2 - 3 | 409 | 95 | |
AK138387.1 | - | 2 - 3 | 409 | 95 | ||
BC049693.1 | - | 3 - 4 | 318 | 95 | ||
BC119515.1 | - | 2 - 3 | 395 | 40055598 | 95 | |
BC119516.1 | - | 2 - 3 | 394 | 40055603 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P234847 | AK005609.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P234847 | AK005609.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720743 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667968 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508009 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676662 | BC006583.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676662 | BC037747.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676662 | XM_485735.5 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676662 | XM_908853.2 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795549 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972181 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819440 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720586 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961024 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910309 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297742 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469584 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669584 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692960 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426532 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155629 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53928 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92288 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12267 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172325 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161002 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164744 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167819 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P234847 | AK005609.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P234847 | AK005609.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164744 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161002 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167819 | - | - |
Whole Mouse Genome G4122A | A_51_P234847 | AK005609.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P234847 | AK005609.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167819 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164744 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110844 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110839 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10750381 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110828 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110834 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110840 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110808 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10529997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110837 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110811 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29110832 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10529993 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10405966 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10405968 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10405967 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10405965 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453643_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10405964 | - | - |
Mouse Genome 430 2.0 Array | 1453643_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453643_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69315_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69315_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300005457.mm.1 | NM_027035.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300005457.mm.1 | NM_027035.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300005457.mm.1 | TC1300000839.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300005458.mm.1 | NM_027035.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300005458.mm.1 | NM_027035.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300005458.mm.1 | TC1300000839.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453643_at | - | - |
Mouse Expression Set 430 | 1453643_PM_at | - | - |
Murine Genome U74v2 Set | 138572_at | - | - |
Murine Genome U74v2 Set | 138572_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453643_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453643_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 350603 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2658067 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2658067 | - | - |
