Assay Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030035.1 | NP_084311.1 | 8 - 9 | 1072 | 72 | |
| GenBank mRNA | BC145221.1 | - | 8 - 9 | 1031 | 9053772 | 72 |
| BC150731.1 | - | 8 - 9 | 1049 | 9087642 | 72 | |
More Information
Set Membership: |
|

Golgb1
Genomic Map