| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300013784.mm.1 |
NM_029636.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013784.mm.1 |
NM_029636.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013784.mm.1 |
TC1300002146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013783.mm.1 |
NM_029636.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013783.mm.1 |
NM_029636.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013783.mm.1 |
TC1300002146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013782.mm.1 |
NM_029636.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013782.mm.1 |
NM_029636.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013782.mm.1 |
TC1300002146.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300013785.mm.1 |
NM_029636.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013785.mm.1 |
NM_029636.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300013785.mm.1 |
TC1300002146.mm.1 |
MTA TC |