Assay Details
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005526.1 | - | 1 - 2 | 71 | 94 | |
AK131661.1 | - | 1 - 2 | 71 | 94 | ||
BC103769.1 | - | 1 - 2 | 60 | 30787117 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK005526.1, AK131661.1, BC103769.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
