Assay Details
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF158182.1 | - | 1 - 2 | 141 | 114 | |
AK005526.1 | - | 2 - 3 | 208 | 114 | ||
AK131661.1 | - | 2 - 3 | 208 | 114 | ||
BC103769.1 | - | 2 - 3 | 197 | 30787117 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
