Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004068.mm.1 |
NM_025329.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004068.mm.1 |
NM_025329.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004068.mm.1 |
TC1600000442.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004070.mm.1 |
NM_025329.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004070.mm.1 |
NM_025329.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004070.mm.1 |
TC1600000442.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004069.mm.1 |
NM_025329.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004069.mm.1 |
NM_025329.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004069.mm.1 |
TC1600000442.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004071.mm.1 |
NM_025329.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004071.mm.1 |
NM_025329.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004071.mm.1 |
TC1600000442.mm.1 |
MTA TC |