Assay Details
Gene Symbol: Tctex1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025329.3 | NP_079605.1 | 2 - 3 | 367 | 92 | |
GenBank mRNA | AK002589.1 | - | 2 - 3 | 326 | 92 | |
AK007178.1 | - | 2 - 3 | 355 | 92 | ||
AK010027.1 | - | 2 - 3 | 325 | 92 | ||
BC059716.1 | - | 2 - 3 | 368 | 6774264 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
