Assay Details
Gene Symbol: Cox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163456.1 | NP_001156928.1 | 3 - 4 | 652 | 75 | |
GenBank mRNA | BC038311.1 | - | 3 - 4 | 422 | 75 | |
BC145479.1 | - | 3 - 4 | 630 | 9054042 | 75 | |
CX225681.1 | - | 3 - 4 | 579 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
