Assay Details
| Gene Symbol: Mthfd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026788.1 | NP_081064.1 | 3 - 4 | 544 | 96 | |
| GenBank mRNA | AK003824.1 | - | 1 - 2 | 111 | 96 | |
| BU610697.1 | - | 2 - 3 | 225 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mthfd2l
Genomic Map