Assay Details
| Gene Symbol: Grk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286066.1 | NP_001272995.1 | 15 - 16 | 1846 | 86 | |
| NM_011938.4 | NP_036068.2 | 15 - 16 | 1853 | 86 | ||
| GenBank mRNA | AF040749.1 | - | 15 - 16 | 1723 | 86 | |
| CA528447.1 | - | 4 - 5 | 452 | 86 | ||
| CD354984.1 | - | 2 - 3 | 193 | 86 | ||
| Y15797.1 | - | 15 - 16 | 1675 | 85 | ||
| Y15800.1 | - | 15 - 16 | 1668 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grk6
Genomic Map