Assay Details
| Gene Symbol: Lipi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252513.1 | NP_001239442.1 | 5 - 6 | 808 | 154 | |
| GenBank mRNA | AK079027.1 | - | 3 - 4 | 219 | 154 | |
| AK086576.1 | - | 5 - 6 | 811 | 154 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK079027.1 transcripts.
Set Membership: |
|

Lipi
Genomic Map