Assay Details
| Gene Symbol: Lipi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252513.1 | NP_001239442.1 | 8 - 9 | 1162 | 110 | |
| GenBank mRNA | AK079027.1 | - | 6 - 7 | 573 | 110 | |
| AK086576.1 | - | 8 - 9 | 112 | |||
| BC147403.1 | - | 3 - 4 | 354 | 9056446 | 110 | |
| BC147404.1 | - | 3 - 4 | 354 | 9056445 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK079027.1, BC147403.1, BC147404.1 transcripts.
Set Membership: |
|

Lipi
Genomic Map