Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300011471.mm.1 |
NM_139063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011471.mm.1 |
NM_139063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011471.mm.1 |
TC1300001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011468.mm.1 |
NM_139063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011468.mm.1 |
NM_139063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011468.mm.1 |
TC1300001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011469.mm.1 |
NM_139063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011469.mm.1 |
NM_139063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011469.mm.1 |
TC1300001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011467.mm.1 |
NM_139063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011467.mm.1 |
NM_139063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011467.mm.1 |
TC1300001836.mm.1 |
MTA TC |