Assay Details
Gene Symbol: Snrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164572.1 | NP_001158044.1 | 3 - 4 | 980 | 59 | |
NM_133741.2 | NP_598502.2 | 4 - 5 | 1030 | 59 | ||
GenBank mRNA | AK082185.1 | - | 1641 | 59 | ||
AK189350.1 | - | 2 - 3 | 208 | 59 | ||
BC020189.1 | - | 4 - 5 | 921 | 4482701 | 59 | |
BC094658.1 | - | 3 - 4 | 907 | 5721174 | 59 | |
BC150742.1 | - | 2 - 3 | 851 | 9087653 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
