Assay Details
| Gene Symbol: 4930553E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040567.1 | 2 - 3 | 198 | 63 | ||
| GenBank mRNA | AK016102.1 | - | 2 - 3 | 199 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neural cell adhesion molecule 2 | Ncam2 |
Ncam-2, Ocam, RNCAM |
-2.7mb | Sense | Chr. 16 - 81200697 - 81624290 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A730009L09 gene | A730009L09Rik | 139.5kb | Sense | Chr. 16 - 84515050 - 84523976 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930553E22Rik
Genomic Map