Assay Details
Gene Symbol: Gm16863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046162.1 | 4 - 5 | 734 | 113 | ||
GenBank mRNA | AK043964.1 | - | 4 - 5 | 735 | 113 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Eph receptor B3 | Ephb3 |
AW456895, Cek10, Etk2, HEK2, MDK5, Sek4, Tyro6 |
-24.2kb | Anti-sense | Chr. 16 - 21204795 - 21223309 on NCBI Build GRCm38 |
Downstream | microRNA 7680 | Mir7680 |
Gm27417, mmu-mir-7680 |
1.5kb | Sense | Chr. 16 - 21256013 - 21256066 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2321919 | AK043964.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287034 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633241 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8282 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494269 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448381 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544725 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389919 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921616 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25488 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175497 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110236 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164959 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132152 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03145139 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08528938 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132152 | - | - |
Whole Mouse Genome G4122A | A_52_P802927 | AK043964.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139105 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591531 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591540 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12653899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591403 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591487 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591507 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591599 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591405 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591511 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591573 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12834555 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12765133 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591509 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12470934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12710173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12903493 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12942480 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591452 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591382 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591634 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12637106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12589751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591535 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591390 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12615968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12572484 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12929900 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591602 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12765127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591533 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13030600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12881034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12710183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591489 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12886142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12912819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591584 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12507325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12470933 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591512 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12470944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12710643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12981078 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591560 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591530 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591544 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12903497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591448 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13001573 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591503 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12470937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591588 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12959872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591447 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591520 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591630 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13020132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12981072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12529943 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591525 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591621 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591538 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591612 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12478590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13001574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591542 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12471507 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591528 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591444 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591490 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591610 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591498 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591505 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13020130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12471510 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591400 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12765126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12881041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12710640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30591618 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1600011304.mm.1 | NR_046162.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600011304.mm.1 | NR_046162.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600011304.mm.1 | TC1600001359.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 700295 | - | - |
