Assay Details
| Gene Symbol: Nr4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038520.1 | - | 1 - 2 | 486 | 148 | |
| BC068150.1 | - | 1 - 2 | 408 | 148 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr4a3| Gene Symbol: Nr4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038520.1 | - | 1 - 2 | 486 | 148 | |
| BC068150.1 | - | 1 - 2 | 408 | 148 | ||
Set Membership: |
|