Assay Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009593.2 | NP_033723.1 | 4 - 5 | 662 | 65 | |
| GenBank mRNA | AF323659.1 | - | 4 - 5 | 658 | 65 | |
| AK037433.1 | - | 4 - 5 | 582 | 65 | ||
| BC119471.2 | - | 4 - 5 | 610 | 40048166 | 65 | |
| U34920.1 | - | 4 - 5 | 620 | 65 | ||
| Z48745.1 | - | 4 - 5 | 649 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcg1
Genomic Map