Assay Details
| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163776.1 | NP_001157248.1 | 5 - 6 | 643 | 149 | |
| NM_080727.2 | NP_542765.2 | 4 - 5 | 503 | 149 | ||
| GenBank mRNA | AK158203.1 | - | 5 - 6 | 646 | 149 | |
| BC112375.1 | - | 4 - 5 | 500 | 40057032 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss3
Genomic Map